Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIF3L1 Monoclonal Antibody (OTI4B5), TrueMAB™, OriGene
SDP

Catalog No. 501674602
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-4602 100 μL
50-254-0702 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-4602 Supplier OriGene Supplier No. TA503798
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALS2CR1 may function as a transcriptional corepressor through its interaction with COPS2, negatively regulating the expression of genes involved in neuronal differentiation.

Specifications

Antigen NIF3L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B5
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NIF3L1
Gene Accession No. Q9GZT8
Gene Alias ALS2CR1, CALS-7, MDS015
Gene Symbols NIF3L1
Host Species Mouse
Immunogen Full length human recombit protein of human NIF3L1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60491
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.