Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIFK Antibody (18E148), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056590AF350
Description
NIFK Monoclonal antibody specifically detects NIFK in Human samples. It is validated for Western BlotSpecifications
NIFK | |
Monoclonal | |
Western Blot | |
hNIFK, MKI67 (FHA domain) interacting nucleolar phosphoprotein, MKI67 FHA domain-interacting nucleolar phosphoprotein, NIFKNOPP34, Nopp34, Nucleolar phosphoprotein Nopp34, Nucleolar protein interacting with the FHA domain of pKI-67 | |
This antibody was developed against a synthetic peptide corresponding to a portion of human NIFK between amino acids 100-200. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
84365 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction