Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIFK Antibody (18E148), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056590G
Description
NIFK Monoclonal specifically detects NIFK in Human samples. It is validated for Western Blot.Specifications
NIFK | |
Monoclonal | |
Western Blot | |
hNIFK, MKI67 (FHA domain) interacting nucleolar phosphoprotein, MKI67 FHA domain-interacting nucleolar phosphoprotein, NIFKNOPP34, Nopp34, Nucleolar phosphoprotein Nopp34, Nucleolar protein interacting with the FHA domain of pKI-67 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
A 34 kDa band is observed. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
DyLight 488 | |
Q9BYG3 | |
MKI67IP | |
This antibody was developed against a synthetic peptide corresponding to a portion of human NIFK between amino acids 100-200. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
84365 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction