Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIFK Antibody (18E148), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$170.97 - $492.89

Specifications

Antigen NIFK
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-3 ug/ml
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056590T
SDP
View Documents
Novus Biologicals
NB10056590SS
0.025 mg
Each for $170.97
Only null left
Add to Cart
 
NB10056590
SDP
View Documents
Novus Biologicals
NB10056590
0.1 mg
Each for $492.89
Only null left
Add to Cart
 
Description

Description

NIFK Monoclonal specifically detects NIFK in Human samples. It is validated for Western Blot.
Specifications

Specifications

NIFK
Western Blot 0.5-3 ug/ml
Monoclonal
Purified
RUO
Human
hNIFK, MKI67 (FHA domain) interacting nucleolar phosphoprotein, MKI67 FHA domain-interacting nucleolar phosphoprotein, NIFKNOPP34, Nopp34, Nucleolar phosphoprotein Nopp34, Nucleolar protein interacting with the FHA domain of pKI-67
MKI67IP
IgG1
Protein G purified
A 34 kDa band is observed.
0.5 mg/mL
Western Blot
Unconjugated
Mouse
Core ESC Like Genes, Stem Cell Markers
Q9BYG3
84365
This antibody was developed against a synthetic peptide corresponding to a portion of human NIFK between amino acids 100-200.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.