Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIFK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056661SS
Description
NIFK Polyclonal specifically detects NIFK in Human, Mouse samples. It is validated for Western Blot.Specifications
NIFK | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
hNIFK, MKI67 (FHA domain) interacting nucleolar phosphoprotein, MKI67 FHA domain-interacting nucleolar phosphoprotein, NIFKNOPP34, Nopp34, Nucleolar phosphoprotein Nopp34, Nucleolar protein interacting with the FHA domain of pKI-67 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
In NIH 3T3, a 34 kDa band is observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:1000 | |
Q9BYG3 | |
MKI67IP | |
This antibody was developed against a synthetic peptide corresponding to amino acids 75-90 of mouse NIFK. | |
0.025 mg | |
Core ESC Like Genes, Stem Cell Markers | |
84365 | |
Human, Mouse | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction