Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIFK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118119
This item is not returnable.
View return policy
Description
NIFK Polyclonal Antibody for Western Blot, ICC/IF, IP
NIFK, also known as nucleolar protein interacting with the forkhead homolog protein, is a gene associated with several cancer types and is considered a potential prognostic biomarker. This gene functions within the nucleolus and plays a critical role in cellular proliferation, metastasis, and invasiveness, particularly noted in hepatocellular carcinoma (HCC) and colorectal cancer. NIFK expression is regulated via several pathways, including the MYC proto-oncogene pathway, and its silencing can lead to decreased fatty acid synthesis in colorectal cancer cells. Furthermore, it's involved in the regulation of innate immunity through pathways such as NF-kB in specific cancer cells. Due to its involvement in cancer progression, NIFK is being explored as a therapeutic target and holds promise for advancing precision medicine strategies across various cancer treatments.
Specifications
NIFK | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9BYG3 | |
Nifk | |
E. coli-derived Human NIFK fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Nifk | |
AI852665; C130020J04Rik; hNIFK; Ki67ip; MKI67 (FHA domain) interacting nucleolar phosphoprotein; MKI67 FHA domain-interacting nucleolar phosphoprotein; MKI67IP; mNIFK; NIFK; NOPP34; nucleolar phosphoprotein Nopp34; nucleolar protein interacting with the FHA domain of MKI67; nucleolar protein interacting with the FHA domain of pKI-67; spermatogenesis-related protein (Srp) | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
84365 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction