Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIFK Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988141
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8141 Supplier Bioss Supplier No. BS7937R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May sustain the structure of the postsynaptic density (PSD).

Specifications

Antigen NIFK
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Nifk
Gene Accession No. Q9BYG3
Gene Alias AI852665; C130020J04Rik; hNIFK; Ki67ip; Mki67 (FHA domain) interacting nucleolar phosphoprotein; MKI67 FHA domain-interacting nucleolar phosphoprotein; Mki67ip; mNIFK; NIFK; NOPP34; Nucleolar phosphoprotein Nopp34; nucleolar protein interacting with the FHA domain of MKI67; Nucleolar protein interacting with the FHA domain of pKI-67
Gene Symbols Nifk
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NIFK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84365
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.