Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nifursol Monoclonal Antibody (3C9)
GREENER_CHOICE

Catalog No. PIMA549671
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549671 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549671 Supplier Invitrogen™ Supplier No. MA549671
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Nifursol is a synthetic antibacterial agent that is used in veterinary medicine to treat a variety of bacterial infections in animals. It belongs to the nitrofuran class of antibiotics and works by inhibiting bacterial growth. Nifursol is commonly used to treat infections in poultry, such as coccidiosis and necrotic enteritis, and has also been used to treat infections in pigs and other animals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nifursol
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3C9
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of NFS hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.