Learn More
MSA™ Ultravue™ Full Facepiece Assembly, Quick Disconnect
Facepiece without nosecup has inhalation check valve and exhalation valve that work together to avoid exhaled air from entering the mask mounted regulator and contaminating the internal regulator components.
Supplier: MSA™ 801448
View more versions of this product
Description
- Pressure-Demand Ultravue
- Full Facepieces
- Rubber hycar material
- Standard 5-point head harness with quick disconnect feature
- Without nosecup
Specifications
NIOSH 42 CFR | |
Facepiece Assembly | |
Speaking Diaphragm | |
Medium |
Pressure-Demand | |
5-strap | |
BMR, Hycar |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.