Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NIH 3T3 Whole Cell Lysate
SDP

Catalog No. NB800PC8 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB800PC8 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB800PC8 Supplier R&D Systems™ Supplier No. NB800PC8

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

The NIH 3T3 Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot, SDS-Page.

Specifications

Quantity 0.1 mL
Storage Buffer Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS diluted in 1X lithium dodecyl sulfate (LDS) sample buffer with bromophenol blue and 40-70% glycerol, pH 8.4. BME has not been added.
Purity Protein
Test Specificity NIH/3T3 whole cell lysate (Positive control)
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.
Product Line Novus
For Use With (Application) Western Blot
Tissue Bone/Cartilage

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.