Learn More
Promega NIK Kinase Enzyme System, 1mg
Catalog No. PRV6453 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6453 1 mg
1 options
Catalog No. PRV6453 Supplier Promega Supplier No. V6453
Only null left
Add to Cart
Add to Cart
Request a Quote
Recombinant human NIK (325-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. NIK is a mitogen-activated protein kinase kinase kinase 14 (MAP3K14), which binds to TRAF2 and stimulates NF-kappaB activity.
  • Easily Screen and Profile NIK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C