Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIK/MAP3K14 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP223603AF647
Description
NIK/MAP3K14 Polyclonal specifically detects NIK/MAP3K14 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NIK/MAP3K14 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Q99558 | |
MAP3K14 | |
A synthetic peptide made to a C-terminal portion of the human MAP3K14 protein (between residues 800-900) [UniProt Q99558] | |
0.1 mL | |
Apoptosis, Cancer, Protein Kinase, Signal Transduction | |
9020 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 2.7.11.25, FTDCR1B, HS, HsNIK, HSNIKserine/threonine protein-kinase, mitogen-activated protein kinase kinase kinase 14, NIKNF-kappa-beta-inducing kinase, Serine/threonine-protein kinase NIK | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human and mouse. Predicted to react with monkey and rat based on sequence homology. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction