Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIK/MAP3K14 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2236030.1ML
Description
NIK/MAP3K14 Polyclonal specifically detects NIK/MAP3K14 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NIK/MAP3K14 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
EC 2.7.11.25, FTDCR1B, HS, HsNIK, HSNIKserine/threonine protein-kinase, mitogen-activated protein kinase kinase kinase 14, NIKNF-kappa-beta-inducing kinase, Serine/threonine-protein kinase NIK | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2.0 mg/mL | |
Western Blot 2.0 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:400 | |
Q99558, Q9WUL6 | |
MAP3K14 | |
A synthetic peptide made to a C-terminal portion of the human MAP3K14 protein (between residues 800-900) [UniProt Q99558] | |
0.1 mL | |
Apoptosis, Cancer, Protein Kinase, Signal Transduction | |
9020 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction