Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIK/MAP3K14 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056367T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056367T 0.025 mg
NB10056367 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056367T Supplier Novus Biologicals Supplier No. NB10056367SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NIK/MAP3K14 Polyclonal specifically detects NIK/MAP3K14 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NIK/MAP3K14
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence 1:100
Formulation PBS with No Preservative
Gene Accession No. Q99558
Gene Alias EC 2.7.11.25, FTDCR1B, HS, HsNIK, HSNIKserine/threonine protein-kinase, mitogen-activated protein kinase kinase kinase 14, NIKNF-kappa-beta-inducing kinase, Serine/threonine-protein kinase NIK
Gene Symbols MAP3K14
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 687-703 of human NIK.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9020
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.