Learn More
Invitrogen™ NIK Polyclonal Antibody

Description
Suggested positive control: antigen standard for MAP3K14 (transient overexpression lysate).
Specifications
Specifications
| Antigen | NIK |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | MAP3K14 |
| Gene Accession No. | Q99558 |
| Gene Alias | aly; alymphoplasia; FTDCR1B; HS; HsNIK; Map3k14; Mitogen-activated protein kinase kinase kinase 14; NF kappa B-inducing kinase; NF-kappa-beta-inducing kinase; Nfkb inducing kinase; NIK; protein kinase; serine/threonine-protein kinase NIK |
| Gene Symbols | MAP3K14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.