Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nile Red Fluorescent stain for intracellular lipid droplets. MP Biomedicals

Catalog No. icn15174450 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
50 mg

Chemical Identifiers

CAS 7385-67-3
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.38
MDL Number MFCD00011639
InChI Key VOFUROIFQGPCGE-UHFFFAOYSA-N
Synonym Nile Blue A Oxazone, 9-diethylamino-5H-benzo-alpha-phenoxazine-5-one
PubChem CID 65182
ChEBI CHEBI:52169
IUPAC Name 9-(diethylamino)benzo[a]phenoxazin-5-one
SMILES CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31

Specifications

Formula Weight 318.4
Quantity 50 mg
Chemical Name or Material Nile Red, Fluorescent stain for intracellular lipid droplets.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.