Learn More
Description
Nimbolide can be used for the development of Degraders that utilize RNF114 as an E3 ligase for Targeted Protein Degradation.
Specifications
Specifications
| Chemical Name or Material | (2 aR,5 aR,6 S,6 aR,8 R,9 aR,10 AS,10 bR,10 cR)-8-(3-Furanyl)-2 A,5 A,6,6 A,8,9,9 A,10 A,10 b,10 c-decahydro-2 A,5 A,6 A,7-tetramethyl-2,5-dioxo-2 H,5 H-cyclopenta[d']naphtho[1,8-bc:2,3-b']difuran-6-acetic acid methyl ester |
| CAS | 25990-37-8 |
| Quantity | 25 mg |
| Target | Ubiquitin Ligase (E3) Inhibitors |
| Molecular Formula | C27H30O7 |
| Purity | 0.97 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

