Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIMP Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8334R

Description
This gene encodes a novel mitochondrial protein that interacts with reticulon 4, which is a potent inhibitor of regeneration following spinal cord injury. The interaction of reticulon 4 with mitochondrial proteins may provide insight into the mechanisms for reticulon-induced inhibition of neurite growth.Specifications
NIMP | |
Polyclonal | |
Unconjugated | |
Rtn4ip1 | |
D10Ertd690e; NIMP; NOGO-interacting mitochondrial protein; OPA10; reticulon 4 interacting protein 1; reticulon 4 interacting protein 1, mitochondrial; Reticulon-4-interacting protein 1, mitochondrial; Rtn4ip1 | |
Rabbit | |
Protein A | |
RUO | |
84816 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8WWV3 | |
Rtn4ip1 | |
KLH conjugated synthetic peptide derived from human NIMP/RTN4IP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction