Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ninein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582224 Supplier Invitrogen™ Supplier No. PA582224
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EERLTQMRNE YERQCRVLQD QVDELQSELE EYRAQGRVLR LPLKNSPSEE VEANSGGIEP EHGLGSEECN PLNMSIEAEL VIEQMKEQHH RDICCLRLEL EDKVRHYEKQ LDETVVSCKK AQENMKQRHE NETHTL Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 74%.

Ninein is a component of subdistal appendages of the mother centriole, localized specifically in the pericentriolar matrix of the centrosome. It may be involved in microtubule minus-end capping, centriole positioning, and protein anchoring. Recent studies have shown that the microtubule nucleation and anchoring at the centrosome are independent processes linked by ninein function. At least five human ninein isoforms with divergent C-terminal domains and two mouse isoforms have been reported. Data indicates that the combined action of domains (C-terminal and N-terminal) might, in the absence of coiled coil region, be sufficient to localize ninein to the mother centriole. Microinjection of antibodies against ninein into metaphase HeLa cells can disrupt the reformation of tubular conformation of proteins within the centrosome following cell division, and consequently lead to dispersal of centrosomal material throughout the cytosol. MTOC (microtubule organizing center) function can be disrupted when anti-ninein antibodies are injected into postmitotic cells. Antibodies against ninein are present in sera from patients with autoimmune diseases that develop autoantibodies to centrosomal proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ninein
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NIN
Gene Accession No. Q8N4C6
Gene Alias 3110068G20Rik; AI385615; AU024711; glycogen synthase kinase 3 beta-interacting protein; GSK3B-interacting protein; hNinein; KIAA1565; mKIAA1565; Nin; Ninein; ninein (GSK3B interacting protein); ninein centrosomal protein; SCKL7
Gene Symbols NIN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NIN. Recombinant protein control fragment (Product #RP-102160).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51199
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.