Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ninein, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1720MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1720MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1720MI Supplier MilliporeSigma™ Supplier No. ABN1720
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ninein (UniProt Q61043; also known as Glycogen synthase kinase 3 beta-interacting protein, GSK3B-interacting protein) is encoded by the Nin (also known as Kiaa1565) gene (Gene ID 18080) in murine species. Ninein is a microtubule (MT) minus-end-binding protein that helps anchor the MTs to the centrosome. Ninein is also found in noncentrosomal locations in association with the minus-end of noncentrosomal MTs present in polarized cells. Ninein is essential for the self-renewal and maintenance of apical progenitors (APs) in developing mouse neocortex by anchoring of MTs to the centrosome, while only noncentrosomal ninein is seen in cortical neurons where it is localized to the axons and the somatodendritic compartment. Centrosome-associated ninein plays an important role in stabilizing the radial MT asters, while noncentrosomal ninein plays an MT-stabilizing role in the axons of neurons. Ninein is a downstream target of the transcription factor Sip1 involved in CNS development. Re-expression of ninein in Sip1-deficient neurons or overexpressing ninein in wild-type neurons significantly increases their resistance to MT depolymerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ninein
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene Nin, Kiaa1565
Gene Accession No. Q61043
Host Species Rabbit
Immunogen His-tagged recombinant mouse ninein fragment corresponding to the region between EF-hands 2&5.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001273008
Test Specificity This polyclonal antiserum targets region between EF-hands 2&5 present in all murine and human ninein spliced isoforms reported by UniProt (Q61043 & Q8N4C6).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.