Alfa Aesar™ Ninhydrin, ACS reagent

Manufacturer: Alfa Aesar™ 4384614

Quantity 25g

 View more versions of this product

Catalog No. AA4384614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 485-47-2
    Molecular Formula C9H6O4
    Formula Weight 178.14
    MDL Number MFCD00003791
    Beilstein 1910963
    Merck Index 14,6554
    Synonym 2,2-Dihydroxy-1,3-indanedione
    Chemical Name or Material Ninhydrin
    Grade ACS Grade

    Specifications

    Formula Weight 178.14
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 485-47-2
    Sensitivity Light Sensitive
    Molecular Formula C9H6O4
    MDL Number MFCD00003791
    Beilstein 1910963
    Merck Index 14,6554
    Health Hazard 1 Warning
    Melting Point 251°C (dec.)
    Color Pale yellow to pale brown
    RTECS Number NK5425000
    Quantity 25g
    TSCA TSCA
    Physical Form Crystalline
    Solubility Information Soluble in water and alcohol
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Synonym 2,2-Dihydroxy-1,3-indanedione
    Chemical Name or Material Ninhydrin
    Grade ACS Grade

    Description

    • Reagent for determination of amino acids and amine containing compounds, as an intermediate

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit