Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninhydrin Solution R, Ricca Chemical
SDP

Catalog No. 530916 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 71-36-3, 7732-18-5, 64-19-7, 485-47-2
Molecular Formula C4H10O
Molecular Weight (g/mol) Mixture
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Min % 92.0, 5.28, 0.71, 0.24
CAS Max % 96.0, 5.5, 0.74, 0.25
Grade Indicator
Packaging Amber Glass Bottle
Color Colorless to Yellow
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ninhydrin Solution, Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.