Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NINJ2 Mouse anti-Human, Clone: 5C5B2, Proteintech
SDP

Catalog No. 501736635 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6635 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6635 Supplier Proteintech Group Inc Supplier No. 662021IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues.

Specifications

Antigen NINJ2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5C5B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NINJ2
Gene Accession No. Q9NZG7
Gene Alias Nerve injury induced protein 2, NINJ2, ninjurin 2
Gene Symbols NINJ2
Host Species Mouse
Immunogen NINJ2 Fusion Protein Ag5343
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4815
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.