Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety™ PredaFlex Shoulder Length Nitrile Glove

Catalog No. 19034387
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large

MCR Safety PredaFlex™ Work Gloves - shoulder length fully coated with nitrile, soft interlock liner, sandy finish.

  • Grommet on the shoulder end for hanging the glove
  • Sandy nitrile finish
  • Shoulder length protection
  • Soft cotton brushed interlock liner
  • Treated with ActiFresh™

Specifications

Accelerator Free No
Applications Industrial
AQL 0.65
Color Yellow
Disposable Reusable
Exterior Finish Smooth
Latex Free No
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size 2X-Large
Sterility Non-sterile
Glove Cuff Style Elasticated
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.