Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ninjurin-1 Antibody (758926), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB51051PECY7
Description
Ninjurin-1 Monoclonal antibody specifically detects Ninjurin-1 in Human samples. It is validated for Flow CytometrySpecifications
Ninjurin-1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4814 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
758926 | |
Flow Cytometry | |
Nerve injury-induced protein 1, nerve injury-induced protein-1, NIN1, NINJURIN, ninjurin 1, ninjurin-1 | |
E. coli-derived recombinant human Ninjurin-1, Asp2-Val81, Accession # Q92982 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction