Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ninjurin 1 Monoclonal Antibody (758926)
GREENER_CHOICE

Catalog No. PIMA524365
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524365 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524365 Supplier Invitrogen™ Supplier No. MA524365
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues. Cell adhesion requires divalent cations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ninjurin 1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 758926
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Ninj1
Gene Accession No. Q92982
Gene Alias AU024536; Nerve injury-induced protein 1; nerve injury-induced protein-1; NIN1; Ninj1; NINJURIN; ninjurin 1; ninjurin-1
Gene Symbols Ninj1
Host Species Mouse
Immunogen E. coli-derived recombinant human Ninjurin-1 Asp2-Val81.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4814
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.