Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NINL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596726 Supplier Invitrogen™ Supplier No. PA596726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDEEENHYVS QLREVYSSCD TTGTGFLDRQ ELTQLCLKLH LEQQLPVLLQ TLLGNDHFAR VNFEEFKEGF VAVLSSNAGV RPSDEDSSSL ESAASSAIPP KYVNGSKWYG RRSRPELCDA ATEARRVPEQ QTQASLKSHL WRSASLESVE SPKSDEEAES TKEAQNELFE AQGQLQTWDS EDFGSPQKSC; Positive Samples: Mouse testis; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center.

Involved in the microtubule organization in interphase cells. Overexpression induces the fragmentation of the Golgi, and causes lysosomes to disperse toward the cell periphery; it also interferes with mitotic spindle assembly. May play a role in ovarian carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NINL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NINL
Gene Accession No. Q6ZQ12
Gene Alias 4930519N13Rik; Gm1004; Gm1634; Kiaa0980; LOW QUALITY PROTEIN: ninein-like protein; mKIAA0980; ninein like; ninein-like; ninein-like protein; NINL; Nlp; RGD1306152
Gene Symbols NINL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human NINL (NP_0794523).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78177
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.