Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nintedanib
GREENER_CHOICESDP

Catalog No. 704950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-495-0 50 mg
70-491-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-495-0 Supplier Tocris Bioscience™ Supplier No. 7049/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent VEGFR, PDGFR and FGFR inhibitor

Nintedanib is a potent inhibitor of vascular endothelial growth factor receptors (VEGFRs), platelet-derived growth factor receptors (PDGFRs) and fibroblast growth factor receptors (FGFRs) (IC50 values are 13 - 34 nM for VEGFR subtypes, 59 - 65 nM for PDGFR subtypes, 37 - 108 nM for FGFR subtypes). Also inhibits Flt-3, Lck, Lyn and Src (IC50 values are 25 - 200 nM). Inhibits proliferation in VEGF-stimulated endothelial cells, and PDGF-stimulated pericytes and vascular smooth muscle cells. Reduces vessel density and integrity, and inhibits tumor growth in a mouse tumor xenograft model.

Specifications

Chemical Name or Material Methyl (Z)-3-[[[4-(N-methyl-2-(4-methylpiperazin-1-yl)acetamido)phenyl)amino)(phenyl)methylene]-2-oxoindoline-6-carboxylate
CAS 656247-17-5
Quantity 50 mg
Molecular Formula C31H33N5O4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.