Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium(IV) oxide, 99+% (metals basis)

Catalog No. p-4408141
Click to view available options
Quantity:
10 g
50 g

CAS: 12034-59-2 | NbO2 | 124.90 g/mol

Niobium(IV) oxide is used as a reducing agent, which reduces carbon dioxide and sulfur dioxide to carbon and sulfur respectively. It finds application in dielectric material for electronic capacitors. It is also used for manufacturing metal niobium metal, in optical glass and in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12034-59-2
Molecular Formula NbO2
Molecular Weight (g/mol) 124.90
MDL Number MFCD00016242
InChI Key HFLAMWCKUFHSAZ-UHFFFAOYSA-N
Synonym niobium dioxide, niobium iv oxide, niobium oxide, columbium dioxide, niobium iv oxide trace metals basis, niobium iv oxide trace metals basis 10g
PubChem CID 82839
IUPAC Name dioxoniobium
SMILES O=[Nb]=O

Specifications

Physical Form Powder
Quantity 10 g
Assay Percent Range >99% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 124.91
Density 5.9 g/mL
Chemical Name or Material Niobium(IV) oxide
EINECSNumber 234-809-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.