Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium(IV) selenide, 99.8% (metals basis)

Catalog No. p-7049167
Click to view available options
Quantity:
10 g
50 g

CAS: 12034-77-4 | NbSe2 | 250.85 g/mol

It is used as a potential substance for thin film solar cells as back contact material. It is widely used in electricity brush & special bearings because of its conductivity and lubricity property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12034-77-4
Molecular Formula NbSe2
Molecular Weight (g/mol) 250.85
MDL Number MFCD00016228
InChI Key CXRFFSKFQFGBOT-UHFFFAOYSA-N
Synonym niobium selenide, columbium selenide, niobium diselenide, diselanylideneniobium, bis selanylidene niobium, niobium diselenide, crystal, niobium diselenide trace metals basis, niobium iv selenide trace metals basis 10g
PubChem CID 82841
IUPAC Name bis(selanylidene)niobium
SMILES [Se]=[Nb]=[Se]

Specifications

Physical Form Powder
Quantity 10 g
UN Number UN3283
Solubility Information Insoluble in water.
Formula Weight 250.83
Percent Purity 99.8%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Niobium(IV) selenide
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
EINECSNumber 234-811-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.