Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium(V) chloride, Puratronic™, 99.999% (metals basis)

Catalog No. p-7058993
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 10026-12-7 | Cl5Nb | 270.156 g/mol

Niobium(V) chloride is the precursor in the preparation of pure niobium, and ferroniobium. It is used as a Lewis acid catalyst in organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-12-7
Molecular Formula Cl5Nb
Molecular Weight (g/mol) 270.156
MDL Number MFCD00011127
InChI Key YHBDIEWMOMLKOO-UHFFFAOYSA-I
Synonym niobium v chloride, niobium pentachloride, niobium chloride nbcl5, nbcl5, columbium pentachloride, niobium v chloride, puratronic, niobium v chloride trace metals basis, niobium v chloride-nb 20-200ppm ta puratrem
PubChem CID 24818
IUPAC Name pentachloroniobium
SMILES Cl[Nb](Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 248.2°C
Melting Point 208.3°C
Physical Form Powder
Quantity 5 g
UN Number UN3260
Merck Index 14,6557
Solubility Information Soluble in alcohol,hydrochloric acid,carbon tetrachloride and chloroform.
Formula Weight 270.2
Percent Purity 99.999%
Grade Puratronic™
Odor Acidic
Sensitivity Moisture sensitive
Density 2.74 g/mL
Assay (metals basis)
Chemical Name or Material Niobium (V) chloride
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 233-059-8
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.