Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium(V) oxalate hydrate

Catalog No. AA4481936
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 1660137-09-6 | C10H12NbO21 | 561.091 g/mol

Niobium(V) oxalate hydrate is used as doping precursors in the PZT composition to improve its piezoelectric properties. It is also used in the preparation of niobium pentoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660137-09-6
MDL Number MFCD00050743
Synonym niobium v oxalate, niobium oxalate
PubChem CID 121233648

Specifications

Physical Form Powder
Quantity 500 g
Molecular Formula C10H5NbO20
InChI Key AXIFGFAGYFPNFC-UHFFFAOYSA-I
SMILES OC(=O)C(=O)O[Nb](OC(=O)C(O)=O)(OC(=O)C(O)=O)(OC(=O)C(O)=O)OC(=O)C(O)=O
IUPAC Name tetrakis[(carboxycarbonyl)oxy]niobio hydrogen oxalate
Molecular Weight (g/mol) 538.04
Formula Weight 538.04 (Anhydrous)
Odor Odorless
Chemical Name or Material Niobium(V) oxalate hydrate
Hazard Category H302+H312
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 244-341-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.