Niobium(V) oxalate hydrate, Alfa Aesar™

Manufacturer: Alfa Aesar 4481914

Quantity 25g

 View more versions of this product

Catalog No. AA4481914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21348-59-4
    Physical Form Powder
    Molecular Formula C10H5NbO20·xH2O
    Formula Weight 538.04
    MDL Number MFCD00050743
    Chemical Name or Material Niobium(V) oxalate hydrate

    Specifications

    Formula Weight 538.04
    Quantity 25g
    EINECS Number 244-341-5
    Physical Form Powder
    Odor Odorless
    Molecular Formula C10H5NbO20·xH2O
    MDL Number MFCD00050743
    CAS 21348-59-4
    Health Hazard 1 H302-H312: Harmful if swallowed. Harmful in contact with skin.
    Chemical Name or Material Niobium(V) oxalate hydrate

    H302-H312: Harmful if swallowed. Harmful in contact with skin.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit