Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium(V) oxide, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
Plastic bottle
1313-96-8
Nb2O5
265.8
MFCD00011128
ZKATWMILCYLAPD-UHFFFAOYSA-N
niobium oxide,niobium v oxide,niobium pentoxide,diniobium pentaoxide,niobia,niobium pentaoxide,diniobium pentoxide,niobium 5+ oxide,nobum oxde
123105
dioxoniobiooxy(dioxo)niobium
O=[Nb](=O)O[Nb](=O)=O
1313-96-8
Nb2O5
265.8
MFCD00011128
ZKATWMILCYLAPD-UHFFFAOYSA-N
niobium oxide,niobium v oxide,niobium pentoxide,diniobium pentaoxide,niobia,niobium pentaoxide,diniobium pentoxide,niobium 5+ oxide,nobum oxde
123105
dioxoniobiooxy(dioxo)niobium
O=[Nb](=O)O[Nb](=O)=O
for ultra trace analysis
CAS Min % 99.95
CAS Max % 100.0
Assay Percent Range 99.99%
Linear Formula Nb2O5
Merck Index 15, 6645
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hf and hot h2so4,soluble in fused potassium hydrogen sulfate or,carbonates or hydroxides of the alkali metals
Formula Weight 265.8
Physical Form Powder
Percent Purity 99.99%
Grade Trace Metal Basis
Density 4.4700g/mL
Packaging Plastic bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in hf and hot h2so4, soluble in fused potassium hydrogen sulfate or, carbonates or hydroxides of the alkali metals
Specific Gravity 4.47
Total Trace Metal Impurities 0.01% max
Color White
Melting Point 1520.0°C
Quantity 250g
Chemical Name or Material Niobium(V) oxide
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 215-213-6