Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NIP |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NIP Polyclonal specifically detects NIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NIP | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
90527 | |
This antibody was developed against a recombinant protein corresponding to amino acids: VAHRMQPHRLKAFFNQSVDEDPMLEWSPEEGGLLSPRYRSMADSPKSQDIPLSEASS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Dual oxidase activator 1, dual oxidase maturation factor 1, dual oxidase maturation factor 1 delta, dual oxidase maturation factor 1 gamma, FLJ32334, homolog of Drosophila Numb-interacting protein, mol, NIPdual oxidase maturation factor 1 alpha, Numb-interacting protein, NUMBIPdual oxidase maturation factor 1 beta | |
DUOXA1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title