Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIP30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584428
Description
Immunogen sequence: KRLKPDPEPD DKNQEPSSCK SLGNTSLSGP SIHCPSAAVC IGILPGLGAY SGSSD.
FAM192A is also known as Nip30. This protein has not bee characterized.
Specifications
NIP30 | |
Polyclonal | |
Unconjugated | |
FAM192A | |
1700001O11Rik; 2310065K24Rik; AI596259; C16orf94; CDA018; CDA10; Fam192a; family with sequence similarity 192 member A; family with sequence similarity 192, member A; NEFA-interacting nuclear protein NIP30; Nip30; PA28G-interacting protein; PIP30; protein FAM192A; PSME3-interacting protein; RGD1307433 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80011 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZU8 | |
FAM192A | |
Recombinant protein corresponding to Human FAM192A. Recombinant protein control fragment (Product #RP-106956). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction