Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115653
Description
Antibody detects endogenous levels of total NIP7.
NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
Specifications
| NIP7 | |
| Polyclonal | |
| Unconjugated | |
| Nip7 | |
| 1110017C15Rik; 60S ribosome subunit biogenesis protein NIP7 homolog; 60S ribosome subunit biogenesis protein NIP-like protein-like protein; 6330509M23Rik; AA408773; AA410017; CGI-37; FLJ10296; HSPC031; HSPC180; hypothetical protein LOC553616; KD93; kDa93; Nip7; NIP7 nucleolar pre-rRNA processing protein; NIP7, nucleolar pre-rRNA processing protein; Nip7p; nuclear import 7; nuclear import 7 homolog; nuclear import 7 homolog (S. cerevisiae); nucleolar pre-rRNA processing protein NIP7; OK/SW-cl.76; OK/SW-cl.78; pEachy; PUA protein; Saccharomyces cerevisiae Nip7p homolog; zgc:110384 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192180, 51388, 66164 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9CXK8, Q9WV50, Q9Y221 | |
| Nip7 | |
| A synthesized peptide derived from human NIP7(Accession Q9Y221), corresponding to amino acid residues Q129-L179. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction