Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565279
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565279 Supplier Invitrogen™ Supplier No. PA565279
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APYAKYKVWIK PGAEQSFLYG NHVLKSGLGR ITENTSQYQG VVVYSMADIP LGFGVAAKST QDCRKVDPMA IVVFHQADIG EYVRHEETLT Highest antigen sequence identity to the following orthologs - mouse 99%, rat 98%.

NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIP7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nip7
Gene Accession No. Q9Y221
Gene Alias 1110017C15Rik; 60S ribosome subunit biogenesis protein NIP7 homolog; 60S ribosome subunit biogenesis protein NIP-like protein-like protein; 6330509M23Rik; AA408773; AA410017; CGI-37; FLJ10296; HSPC031; HSPC180; hypothetical protein LOC553616; KD93; kDa93; Nip7; NIP7 nucleolar pre-rRNA processing protein; NIP7, nucleolar pre-rRNA processing protein; Nip7p; nuclear import 7; nuclear import 7 homolog; nuclear import 7 homolog (S. cerevisiae); nucleolar pre-rRNA processing protein NIP7; OK/SW-cl.76; OK/SW-cl.78; pEachy; PUA protein; Saccharomyces cerevisiae Nip7p homolog; zgc:110384
Gene Symbols Nip7
Host Species Rabbit
Immunogen Recombinant Human NIP7. Recombinant protein control fragment (Product #RP-104724).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51388
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.