Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIPA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554954
Description
Immunogen sequence: IQVHVTACIL SVCGWACSSS LESMQLSLIT CSQCMRKVGL WGFQQIESSM TDLDASFGLT SSPIPGLEGR PERLPLVPES PRRMMTRSQD Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
This gene encodes an F-box-containing protein that is a component of an SCF-type E3 ubiquitin ligase complex that regulates the onset of cell division. The G2/M transition in the cell cycle requires the interaction of the proteins cyclin B1 and cyclin-dependent kinase 1. The activated ubiquitin ligase complex targets the protein cyclin B1 for degradation, preventing this transition to mitosis.
Specifications
NIPA | |
Polyclonal | |
Unconjugated | |
ZC3HC1 | |
1110054L24Rik; AU018540; AU019789; C3HC type 1; hematopoietic stem/progenitor cell protein 216; hNIPA; HSPC216; mNIPA; Nipa; nuclear interacting partner of ALK; nuclear interacting partner of anaplastic lymphoma kinase (Alk); nuclear-interacting partner of ALK; Nuclear-interacting partner of anaplastic lymphoma kinase; OTTHUMP00000212330; RGD1561099; ZC3HC1; zinc finger C3HC-type containing 1; zinc finger C3HC-type protein 1; zinc finger, C3HC type 1; zinc finger, C3HC-type 1; zinc finger, C3HC-type containing 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
232679, 296957, 51530 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q80YV2, Q86WB0 | |
ZC3HC1 | |
Recombinant protein corresponding to Human NIPA. Recombinant protein control fragment (Product #RP-92633). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction