Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIPA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582664
Description
Immunogen sequence: CEGQAFAVGV EKNWGAVVRS PEGTPQKIRQ LIDEGIAPEE GGVDAKDTSA TSQSVNGSPQ AEQPSLESTS KEAFFSRVET FSSLKWAGKP FELSPLVCAK YGWV.
This gene encodes an F-box-containing protein that is a component of an SCF-type E3 ubiquitin ligase complex that regulates the onset of cell division. The G2/M transition in the cell cycle requires the interaction of the proteins cyclin B1 and cyclin-dependent kinase 1. The activated ubiquitin ligase complex targets the protein cyclin B1 for degradation, preventing this transition to mitosis.
Specifications
NIPA | |
Polyclonal | |
Unconjugated | |
ZC3HC1 | |
1110054L24Rik; AU018540; AU019789; C3HC type 1; hematopoietic stem/progenitor cell protein 216; hNIPA; HSPC216; mNIPA; Nipa; nuclear interacting partner of ALK; nuclear interacting partner of anaplastic lymphoma kinase (Alk); nuclear-interacting partner of ALK; Nuclear-interacting partner of anaplastic lymphoma kinase; OTTHUMP00000212330; RGD1561099; ZC3HC1; zinc finger C3HC-type containing 1; zinc finger C3HC-type protein 1; zinc finger, C3HC type 1; zinc finger, C3HC-type 1; zinc finger, C3HC-type containing 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51530 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WB0 | |
ZC3HC1 | |
Recombinant protein corresponding to Human ZC3HC1. Recombinant protein control fragment (Product #RP-92632). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction