Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598587 Supplier Invitrogen™ Supplier No. PA598587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NIPA Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes an F-box-containing protein that is a component of an SCF-type E3 ubiquitin ligase complex that regulates the onset of cell division. The G2/M transition in the cell cycle requires the interaction of the proteins cyclin B1 and cyclin-dependent kinase 1. The activated ubiquitin ligase complex targets the protein cyclin B1 for degradation, preventing this transition to mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIPA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZC3HC1
Gene Accession No. Q86WB0
Gene Alias 1110054L24Rik; AU018540; AU019789; C3HC type 1; hematopoietic stem/progenitor cell protein 216; hNIPA; HSPC216; mNIPA; Nipa; nuclear interacting partner of ALK; nuclear interacting partner of anaplastic lymphoma kinase (Alk); nuclear-interacting partner of ALK; Nuclear-interacting partner of anaplastic lymphoma kinase; OTTHUMP00000212330; RGD1561099; ZC3HC1; zinc finger C3HC-type containing 1; zinc finger C3HC-type protein 1; zinc finger, C3HC type 1; zinc finger, C3HC-type 1; zinc finger, C3HC-type containing 1
Gene Symbols ZC3HC1
Host Species Rabbit
Immunogen Recombinant Human Nuclear-interacting partner of ALK protein (1-260AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51530
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.