Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIPA4 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577557
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
NIPAL4 acts as a Mg(2+) transporter and can also transport other divalent cations such as Ba(2+), Mn(2+), Sr(2+) and Co(2+) but to a much less extent than Mg(2+). NIPAL4 may also be a receptor for ligands (trioxilins A3 and B3) from the hepoxilin pathway. The NIPAL4 gene likely encodes a membrane receptor. Mutations in the NIPAL4 gene have been associated with autosomal recessive congenital ichthyosis. Diseases associated with NIPAL4 include Ichthyosis, Congenital, Autosomal Recessive 6 and Autosomal Recessive Congenital Ichthyosis.
Specifications
NIPA4 (extracellular) | |
Polyclonal | |
Unconjugated | |
NIPAL4 | |
Magnesium Transporter NIPA4 (extracellular) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
214112, 303070, 348938 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunofluorescence, Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q0D2K0, Q8BZF2 | |
NIPAL4 | |
Peptide (C)EKVTTIMEMASKMKD, corresponding to amino acid residues 242-256 of human Magnesium Transporter NIPA4 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction