Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIPA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558315 Supplier Invitrogen™ Supplier No. PA558315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKDLDISCAS LPHMHKNPPP SPAPEPTVIR LEDKNVLVDN IELASTSSPE EKPKVFIIHS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 82%.

NIPAL4 acts as a Mg(2+) transporter and can also transport other divalent cations such as Ba(2+), Mn(2+), Sr(2+) and Co(2+) but to a much less extent than Mg(2+). NIPAL4 may also be a receptor for ligands (trioxilins A3 and B3) from the hepoxilin pathway. The NIPAL4 gene likely encodes a membrane receptor. Mutations in the NIPAL4 gene have been associated with autosomal recessive congenital ichthyosis. Diseases associated with NIPAL4 include Ichthyosis, Congenital, Autosomal Recessive 6 and Autosomal Recessive Congenital Ichthyosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIPA4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NIPAL4
Gene Accession No. Q0D2K0
Gene Alias 9530066K23Rik; ARCI6; ICHN; Ichthyin; ICHYN; Magnesium transporter NIPA4; NIPA like domain containing 4; Nipa4; NIPAL4; NIPA-like domain containing 4; NIPA-like protein 4; Non-imprinted in Prader-Willi/Angelman syndrome region protein 4; non-imprinted in Prader-Willi/Angelman syndrome region protein 4 homolog; RGD1309452
Gene Symbols NIPAL4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NIPA4. Recombinant protein control fragment (Product #RP-96939).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.