Learn More
Invitrogen™ Nipah Virus Nucleoprotein Recombinant Rabbit Monoclonal Antibody (HL1436)

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | Nipah Virus Nucleoprotein |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | HL1436 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | Nipah NP |
| Host Species | Rabbit |
| Immunogen | Full length Nipah virus Nucleoprotein recombinant protein. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.