Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIPAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557803 Supplier Invitrogen™ Supplier No. PA557803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTDITWSELT STAKKEAVSL NVNENNYVLL ENLECSAPGY NDDVTLFSRT DD Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 69%.

Acts as a Mg(2+) transporter. Can also transport other divalent cations such as Fe(2+), Sr(2+), Ba(2+), Mn(2+), Cu(2+) and Co(2+) but to a much less extent than Mg(2+).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIPAL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NIPAL1
Gene Accession No. Q6NVV3
Gene Alias 3830408G10Rik; Magnesium transporter NIPA3; NIPA like domain containing 1; Nipa3; Nipal1; NIPA-like domain containing 1; NIPA-like protein 1; Non-imprinted in Prader-Willi/Angelman syndrome region protein 3; non-imprinted in Prader-Willi/Angelman syndrome region protein 3 homolog; Npal1
Gene Symbols NIPAL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NIPAL1. Recombinant protein control fragment (Product #RP-96706).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152519
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.