Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIPBL Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093320AF700
Description
NIPBL Polyclonal antibody specifically detects NIPBL in Human samples. It is validated for Western BlotSpecifications
NIPBL | |
Polyclonal | |
Western Blot | |
CDLS1, delangin, DKFZp434L1319, FLJ11203, FLJ12597, FLJ13354, FLJ13648, FLJ44854, IDN3-B, IDN3CDLS, Nipped-B homolog (Drosophila), nipped-B-like protein, Scc2, SCC2 homolog, sister chromatid cohesion 2 homolog | |
The immunogen recognized by this antibody maps to a region between residue 1025 and 1075 of human Nipped-B-like using the numbering given in entry NP_597677.2 (GeneID 25836). | |
0.1 mL | |
Cell Cycle and Replication | |
25836 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction