Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIPBL Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP177974
Description
NIPBL Polyclonal specifically detects NIPBL in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
NIPBL | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
CDLS1, delangin, DKFZp434L1319, FLJ11203, FLJ12597, FLJ13354, FLJ13648, FLJ44854, IDN3-B, IDN3CDLS, Nipped-B homolog (Drosophila), nipped-B-like protein, Scc2, SCC2 homolog, sister chromatid cohesion 2 homolog | |
Goat | |
316.05 kDa | |
0.1 mg | |
Cell Cycle and Replication | |
25836 | |
Human, Mouse, Rat, Bovine, Canine, Orangutan | |
IgG |
Western Blot, ELISA | |
1.6 mg/ml | |
Western Blot 1:500-1:3000, ELISA 1:2000-1:10000 | |
Q6KC79 | |
NIPBL | |
This affinity-purified NIPBL Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acids 330-365 of Human NIPBL. (Uniprot: Q6KC79) | |
Affinity Purified | |
RUO | |
Primary | |
This affinity purified antibody is directed against human NIPBL protein. A BLAST analysis was used to suggest reactivity with this protein from human, rat, mouse, dog, bovine and orangutan based on 100% homology for the immunogen sequence. Expect reactivity with multiple splice variants of this protein. Cross reactivity with NIPBL homologues from other sources has not been determined. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction