Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NIPBL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113033
Description
NIPBL Polyclonal Antibody for Western Blot, IHC (P), ELISA
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.
Specifications
| NIPBL | |
| Polyclonal | |
| Unconjugated | |
| NIPBL | |
| CDLS; CDLS1; delangin; Delangin homolog; DKFZp434L1319; FLJ11203; FLJ12597; FLJ13354; FLJ13648; Idn3; IDN3-B; NIPBL; NIPBL, cohesin loading factor; Nipped-B homolog; Nipped-B homolog (Drosophila); nipped-B-like protein; SCC2; SCC2 homolog; sister chromatid cohesion 2 homolog | |
| Rabbit | |
| Protein G | |
| RUO | |
| 25836 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6KC79 | |
| NIPBL | |
| Recombinant Human Nipped-B-like protein (2523-2697AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction