Learn More
NIPBL Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301779A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Probably plays a structural role in chromatin. Involved in sister chromatid cohesion, possibly by interacting with the cohesin complex.Specifications
| NIPBL | |
| Polyclonal | |
| Unconjugated | |
| NIPBL | |
| CDLS, CDLS1, delangin, DKFZp434L1319, FLJ11203, FLJ12597, FLJ13354, FLJ13648, IDN3, IDN3-B, Nipped-B homolog, Scc2, SCC2 homolog, sister chromatid cohesion 2 homolog, sister chromatid cohesion 2 homolog (yeast) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25836 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q6KC79 | |
| NIPBL | |
| Between 1025 and 1075 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.