Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIPSNAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114241 Supplier Invitrogen™ Supplier No. PA5114241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRTYKLKPGT MIEWGNNWAR AIKYRQENQE AVGGFFSQIG ELYVVHHLWA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

NIPSNAP is a member of an evolutionarily well conserved gene family and has a strong sequence similarity to the central portion of a protein encoded by C. elegans chromosome III between a 4-nitrophenylphosphatase (NIP) domain and non-neuronal SNAP25-like protein. Recent studies have indicated that NIPSNAP is involved in the regulation of the Ca2+-selective transient receptor potential vanilloid channel 6 (TRPV6). NIPSNAP1 associates with TRPV6 at the plasma membrane and inhibits TRPV6 currents. Other studies show that NIPSNAP's expression is reduced in the phenylketonuria (PKU) mouse brain, suggesting that NIPSNAP may play a role in memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIPSNAP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NIPSNAP1
Gene Accession No. Q9BPW8
Gene Alias 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 1; 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1; 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1 (C. elegans); nipsnap homolog 1 (C. elegans); Nipsnap1; Protein NipSnap homolog 1
Gene Symbols NIPSNAP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human NIPSNAP1 (aa 187-236).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8508
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.